missing translation for 'onlineSavingsMsg'
Learn More

Cholesteryl nonanoate, Thermo Scientific Chemicals

Product Code. 11367875
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11367875 100 g 100g
11357875 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11367875 Supplier Thermo Scientific Alfa Aesar Supplier No. L02857.22
 Restricted Item
Estimated Shipment: 04-09-2026
Add to basket
This item is not returnable. View return policy

Cholesteryl nonanoate is used in hair colors, cosmetic products like lip products, lotions. It is also used in the manufature of some pleochroic dyes. It can be also used as a component of the liquid crystals used for liquid crystal displays.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cholesteryl nonanoate is used in hair colors, cosmetic products like lip products, lotions. It is also used in the manufature of some pleochroic dyes. It can be also used as a component of the liquid crystals used for liquid crystal displays.

Solubility
Soluble in Chloroform, Tetrahydrofuran(THF). Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible materials are Strong oxidizing agents, Strong acids, Strong bases, Strong reducing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1182-66-7
Molecular Formula C36H62O2
Molecular Weight (g/mol) 526.89
MDL Number MFCD00003643
InChI Key WCLNGBQPTVENHV-MKQVXYPISA-N
Synonym cholesteryl pelargonate, cholesteryl nonanoate, cholesterol pelargonate, unii-4313o7p4xw, 5-cholesten-3beta-ol 3-nonanoate, cholesterol, nonanoate, 3beta-hydroxy-5-cholestene 3-nonanoate, nonanoic acid cholesterol ester, pelargonic acid cholesterol ester, cholest-5-ene-3-beta-yl nonanoate
PubChem CID 2723614
IUPAC Name [(3S,8S,9S,10R,13R,14S,17R)-10,13-dimethyl-17-[(2R)-6-methylheptan-2-yl]-2,3,4,7,8,9,11,12,14,15,16,17-dodecahydro-1H-cyclopenta[a]phenanthren-3-yl] nonanoate
SMILES CCCCCCCCC(=O)OC1CCC2(C3CCC4(C(C3CC=C2C1)CCC4C(C)CCCC(C)C)C)C

Specifications

Melting Point 75°C to 79°C
Odor Odorless
Quantity 100 g
Beilstein 3179820
Solubility Information Soluble in Chloroform,Tetrahydrofuran(THF). Insoluble in water.
Optical Rotation −30° (c=5 in Chloroform)
Formula Weight 526.89
Chemical Name or Material Cholesteryl nonanoate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.