missing translation for 'onlineSavingsMsg'
Learn More

Cholesteryl pelargonate, +99%, Thermo Scientific Chemicals

Product Code. 10579843 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10579843 100 g Glass bottle 100g
10550011 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10579843 Supplier Thermo Scientific Acros Supplier No. 110271000
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1182-66-7
Molecular Formula C36H62O2
Molecular Weight (g/mol) 526.89
MDL Number MFCD00003643
InChI Key WCLNGBQPTVENHV-MKQVXYPISA-N
Synonym cholesteryl pelargonate, cholesteryl nonanoate, cholesterol pelargonate, unii-4313o7p4xw, 5-cholesten-3beta-ol 3-nonanoate, cholesterol, nonanoate, 3beta-hydroxy-5-cholestene 3-nonanoate, nonanoic acid cholesterol ester, pelargonic acid cholesterol ester, cholest-5-ene-3-beta-yl nonanoate
PubChem CID 2723614
IUPAC Name [(3S,8S,9S,10R,13R,14S,17R)-10,13-dimethyl-17-[(2R)-6-methylheptan-2-yl]-2,3,4,7,8,9,11,12,14,15,16,17-dodecahydro-1H-cyclopenta[a]phenanthren-3-yl] nonanoate
SMILES CCCCCCCCC(=O)OC1CCC2(C3CCC4(C(C3CC=C2C1)CCC4C(C)CCCC(C)C)C)C

Specifications

Color White
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Glass bottle
Quantity 100 g
Specific Rotation Condition −30.00 (25.00°C c=2,CHCl3)
Formula Weight 526.88
Percent Purity ≥99.0%
Physical Form Powder
Chemical Name or Material Cholesteryl pelargonate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.