missing translation for 'onlineSavingsMsg'
Learn More

Cholic acid, MP Biomedicals™

Product Code. 11453320 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
1 kg
Unit Size:
1kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11453320 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11453320 Supplier MP Biomedicals Supplier No. 0210138391
 Restricted Item
Estimated Shipment: 29-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 81-25-4
Color White
Assay Percent Range 99.6%
Molecular Formula C24H40O5
MDL Number MFCD00003672
Quantity 1 kg
Synonym cholic acid, cholalic acid, cholate, cholalin, colalin, cholsaeure, cholbam, cholicacid, cholic acid, 5beta, 3alpha,7alpha,12alpha-trihydroxy-5beta-cholanic acid
InChI Key BHQCQFFYRZLCQQ-OELDTZBJSA-N
SMILES CC(CCC(=O)O)C1CCC2C1(C(CC3C2C(CC4C3(CCC(C4)O)C)O)O)C
IUPAC Name (4R)-4-[(3R,5S,7R,8R,9S,10S,12S,13R,14S,17R)-3,7,12-trihydroxy-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl]pentanoic acid
Molecular Weight (g/mol) 408.579
PubChem CID 221493
ChEBI CHEBI:16359
Formula Weight 408.58
Physical Form Powder
Chemical Name or Material Cholic acid, from Bovine or Ovine Bile
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.