missing translation for 'onlineSavingsMsg'
Learn More

Cholic Acid, TRC

Product Code. 17993935
Change view
Click to view available options
Quantity:
5 g
25 g
50 g
Unit Size:
25g
50g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17993935 50 g 50g
17990745 25 g 25g
17973835 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17993935 Supplier Toronto Research Chemicals Supplier No. TRCC43260050G
 Restricted Item
Estimated Shipment: 30-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 81-25-4
Molecular Formula C24 H40 O5
Molecular Weight (g/mol) 408.57
Synonym Cholic Acid, (3α,5β,7α,12α)-3,7,12-Trihydroxycholan-24-oic acid, Cholic acid, 17β-[1-Methyl-3-carboxypropyl]etiocholane-3α,7α,12α-triol, 3α,7α,12α-Trihydroxy-5β-cholan-24-oic acid, 3α,7α,12α-Trihydroxy-5β-cholanic acid, 3α,7α,12α-Trihydroxy-5β-cholanoic acid, 3α,7α,12α-Trihydroxy-β-cholanic acid, 3α,7α,12α-Trihydroxycholanic acid, 5β-Cholanic acid-3α,7α,12α-triol, 5β-Cholic acid, Cholalic acid, Cholalin, Colalin, E 1000, NSC-6135, Cholan-24-oic acid, 3,7,12-trihydroxy-, (3α,5β,7α,12α)-
IUPAC Name (4R)-4-[(3R,5S,7R,8R,9S,10S,12S,13R,14S,17R)-3,7,12-trihydroxy-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl]pentanoic acid
SMILES C[C@H](CCC(=O)O)[C@H]1CC[C@H]2[C@@H]3[C@H](O)C[C@@H]4C[C@H](O)CC[C@]4(C)[C@H]3C[C@H](O)[C@]12C

Specifications

Quantity 50 g
InChI Formula InChI=1S/C24H40O5/c1-13(4-7-21(28)29)16-5-6-17-22-18(12-20(27)24(16,17)3)23(2)9-8-15(25)10-14(23)11-19(22)26/h13-20,22,25-27H,4-12H2,1-3H3,(H,28,29)/t13-,14+,15-,16-,17+,18+,19-,20+,22+,23+,24-/m1/s1
Formula Weight 408.2876
Percent Purity >95%
Chemical Name or Material Cholic Acid
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.