missing translation for 'onlineSavingsMsg'
Learn More

Choline bitartrate, 98+%

Product Code. 15436509
Change view
Click to view available options
Quantity:
250 g
1 kg
2.5 kg
Unit Size:
1kg
2.5kg
250g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15436509 250 g 250g
15446509 1 kg 1kg
15456509 2.5 kg 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15436509 Supplier Thermo Scientific Alfa Aesar Supplier No. J64657.30
 Restricted Item
In Stock
Add to basket
Edge

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 87-67-2 | C9H19NO7 | 253.251 g/mol

Choline Bitartrate is used by nerve cells to manufacture a chemical called acetylcholine, which effectively allows cells to communicate with each other. Choline is also heavily associated with neurological function and can help improve movement, coordination, and myofibril contraction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Choline Bitartrate is used by nerve cells to manufacture a chemical called acetylcholine, which effectively allows cells to communicate with each other. Choline is also heavily associated with neurological function and can help improve movement, coordination, and myofibril contraction.

Solubility
Soluble in alcohol and slightly soluble in water.

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 87-67-2
Molecular Formula C9H19NO7
Molecular Weight (g/mol) 253.251
MDL Number MFCD00036332
InChI Key QWJSAWXRUVVRLH-UHFFFAOYSA-M
Synonym choline bitartrate, cholini bitartras, choline 3-carboxy-2,3-dihydroxypropanoate, 2-hydroxyethyl trimethylammonium bitartrate, choline tartrate 1:1, 2-hydroxy-n,n,n-trimethylethanaminium 3-carboxy-2,3-dihydroxypropanoate, cholinibitatratis, choline hydrogen l-+-tartrate, ethanaminium, 2-hydroxy-n,n,n-trimethyl-, 2r,3r-2,3-dihydroxybutanedioate 1:1
PubChem CID 10198924
IUPAC Name 2-hydroxyethyl(trimethyl)azanium;2,3,4-trihydroxy-4-oxobutanoate
SMILES C[N+](C)(C)CCO.C(C(C(=O)[O-])O)(C(=O)O)O

Specifications

Melting Point 151°C to 153°C
Color White
Quantity 250 g
Sensitivity Hygroscopic
Solubility Information Soluble in alcohol and slightly soluble in water.
Formula Weight 253.25
Percent Purity ≥98%
Physical Form Powder
Chemical Name or Material Choline bitartrate
compliance-icons
Product Identifier
  • Choline bitartrate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Choline bitartrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.