missing translation for 'onlineSavingsMsg'
Learn More

Choline bitatrate, >98%, MP Biomedicals™

Product Code. 11463320 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
25 g
250 g
500 g
Unit Size:
250g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11463320 25 g 25g
11473320 250 g 250g
11483320 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11463320 Supplier MP Biomedicals Supplier No. 0210138425
 Restricted Item
Estimated Shipment: 18-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 87-67-2
Molecular Formula C9H19NO7
Molecular Weight (g/mol) 253.251
MDL Number MFCD00036332
InChI Key QWJSAWXRUVVRLH-UHFFFAOYSA-M
Synonym choline bitartrate, cholini bitartras, choline 3-carboxy-2,3-dihydroxypropanoate, 2-hydroxyethyl trimethylammonium bitartrate, choline tartrate 1:1, 2-hydroxy-n,n,n-trimethylethanaminium 3-carboxy-2,3-dihydroxypropanoate, cholinibitatratis, choline hydrogen l-+-tartrate, ethanaminium, 2-hydroxy-n,n,n-trimethyl-, 2r,3r-2,3-dihydroxybutanedioate 1:1
PubChem CID 10198924
IUPAC Name 2-hydroxyethyl(trimethyl)azanium;2,3,4-trihydroxy-4-oxobutanoate
SMILES C[N+](C)(C)CCO.C(C(C(=O)[O-])O)(C(=O)O)O

Specifications

Color White
Assay Percent Range >98%
Quantity 25 g
Formula Weight 253.3
Physical Form Powder
Chemical Name or Material Choline bitatrate, Molecular biology; Cell biology
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.