missing translation for 'onlineSavingsMsg'
Learn More

Choline chloride, 98+% (dry basis)

Product Code. 15815268 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
5 lb.
10 kg
Packaging:
Plastic bottle
Unit Size:
10kg
2268g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15815268 10 kg Plastic bottle 10kg
15825108 5 lb. Plastic bottle 2268g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15815268 Supplier Thermo Scientific Supplier No. J13410.A9
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 67-48-1 | C5H14ClNO | 139.62 g/mol

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67-48-1
Molecular Formula C5H14ClNO
Molecular Weight (g/mol) 139.62
MDL Number MFCD00011721
InChI Key SGMZJAMFUVOLNK-UHFFFAOYSA-M
Synonym choline chloride, hepacholine, biocolina, biocoline, lipotril, paresan, luridin chloride, choline hydrochloride, cholinium chloride, bilineurin chloride
PubChem CID 6209
ChEBI CHEBI:133341
SMILES [Cl-].C[N+](C)(C)CCO

Specifications

Melting Point ∼305°C (decomposition)
Packaging Plastic bottle
Quantity 10 kg
Beilstein 3563126
IUPAC Name (2-hydroxyethyl)trimethylazanium chloride
Formula Weight 139.62
Percent Purity ≥98 (dry basis)
Physical Form Crystalline Powder
Chemical Name or Material Choline chloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.