missing translation for 'onlineSavingsMsg'
Learn More

Choline chloride, 99%, Cell culture reagent grade, MP Biomedicals™

Product Code. 11465102 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
1 kg
Unit Size:
1kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11465102 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11465102 Supplier MP Biomedicals Supplier No. 0219463991
 Restricted Item
Estimated Shipment: 20-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Cell culture grade organic compound used as a substrate for choline acetyltransferase and a source of methyl groups.

Specifications

CAS 67-48-1
Molecular Formula C5H14ClNO
MDL Number MFCD00011721
Quantity 1 kg
Synonym choline chloride, hepacholine, biocolina, biocoline, lipotril, paresan, luridin chloride, choline hydrochloride, bilineurin chloride, 2-hydroxyethyl trimethylammonium chloride
InChI Key SGMZJAMFUVOLNK-UHFFFAOYSA-M
SMILES [Cl-].C[N+](C)(C)CCO
IUPAC Name (2-hydroxyethyl)trimethylazanium chloride
Molecular Weight (g/mol) 139.62
PubChem CID 6209
ChEBI CHEBI:133341
Percent Purity >99%
Grade Cell Culture
Physical Form Crystalline Powder
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.