missing translation for 'onlineSavingsMsg'
Learn More

Choline Chloride, 99%

Product Code. 10703924 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
50 g
500 g
1 kg
5 kg
Packaging:
Plastic bottle
Plastic drum
Unit Size:
1kg
500g
50g
5kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10703924 5 kg Plastic drum 5kg
10011973 50 g Plastic bottle 50g
10570181 500 g Plastic bottle 500g
10715561 1 kg Plastic bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10703924 Supplier Thermo Scientific Acros Supplier No. 110290050
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 67-48-1 | C5H14ClNO | 139.62 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67-48-1
Molecular Formula C5H14ClNO
Molecular Weight (g/mol) 139.62
MDL Number MFCD00011721
InChI Key SGMZJAMFUVOLNK-UHFFFAOYSA-M
Synonym choline chloride, hepacholine, biocolina, biocoline, lipotril, paresan, luridin chloride, choline hydrochloride, cholinium chloride, bilineurin chloride
PubChem CID 6209
ChEBI CHEBI:133341
IUPAC Name 2-hydroxyethyl(trimethyl)azanium;chloride
SMILES [Cl-].C[N+](C)(C)CCO

Specifications

Melting Point 298.0°C to 304.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98.5 to 101.5% (on dry substance) (Argentometry)
Packaging Plastic drum
Linear Formula HOCH2CH2N(CH3)3Cl
Quantity 5 kg
Beilstein 04, II, 722
Merck Index 15, 2211
Solubility Information Solubility in water: soluble. Other solubilities: soluble in ethanol
Formula Weight 139.62
Percent Purity 99%
Physical Form Adhering Crystals
Chemical Name or Material Choline chloride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.