missing translation for 'onlineSavingsMsg'
Learn More

Choline Chloride, BAKER ANALYZED™ Reagent, J.T. Baker™

Product Code. 15547914 Shop All J.T.Baker Products
Change view
Click to view available options
Quantity:
500 g
Packaging:
Poly Bottle
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15547914 500 g Poly Bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15547914 Supplier Avantor J.T.Baker Supplier No. 158201

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High quality chemicals for laboratory and specialized industrial use

Specifications

CAS 67-48-1
Packaging Poly Bottle
Molecular Formula C5H14ClNO
MDL Number MFCD00011721
Quantity 500 g
Synonym choline chloride, hepacholine, biocolina, biocoline, lipotril, paresan, luridin chloride, choline hydrochloride, cholinium chloride, bilineurin chloride
InChI Key SGMZJAMFUVOLNK-UHFFFAOYSA-M
SMILES [Cl-].C[N+](C)(C)CCO
IUPAC Name 2-hydroxyethyl(trimethyl)azanium;chloride
Molecular Weight (g/mol) 139.62
PubChem CID 6209
ChEBI CHEBI:133341
Formula Weight 139.63
Percent Purity 98.0 to 100.5%
Grade BAKER ANALYZED™ Reagent
Chemical Name or Material Choline Chloride
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.