missing translation for 'onlineSavingsMsg'
Learn More

Choline Chloride, TRC

Product Code. 30010149
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30010149 5 g 5g
30010735 25 g 25g
17929984 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30010149 Supplier Toronto Research Chemicals Supplier No. TRCC4326505G
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67-48-1
Molecular Formula C5 H14 N O . Cl
Molecular Weight (g/mol) 139.62
Synonym Choline, chloride (8CI), Ethanaminium, 2-hydroxy-N,N,N-trimethyl-, chloride (9CI), (2-Hydroxyethyl)trimethylammonium chloride, (β-Hydroxyethyl)trimethylammonium chloride, Actiron CC 6, Bilineurin chloride, Biocolina, Biocoline, COL 24, Choline chloride, Cholinium chloride, Claytreat, Hepacholine, Hormocline, Lipotril, Luridin chloride, Neocolina, Norcol 25, Paresan, ReaShure Choline, Trimethyl(2-hydroxyethyl)ammonium chloride
IUPAC Name 2-hydroxyethyl(trimethyl)azanium;chloride
SMILES [Cl-].C[N+](C)(C)CCO

Specifications

Quantity 5 g
InChI Formula InChI=1S/C5H14NO.ClH/c1-6(2,3)4-5-7;/h7H,4-5H2,1-3H3;1H/q+1;/p-1
Formula Weight 139.0764
Percent Purity >95%
Chemical Name or Material Choline Chloride
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.