missing translation for 'onlineSavingsMsg'
Learn More

Choline hydroxide, 46% w/w aq. soln.

Product Code. 11319859
Click to view available options
Quantity:
25 mL
100 mL
500 mL
Unit Size:
100mL
25mL
500mL
This item is not returnable. View return policy

Product Code. 11319859

Brand: Thermo Scientific Alfa Aesar B24443.AC

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 123-41-1 | C5H15NO2 | 121.18 g/mol

Choline hydroxide is used as a precursor molecule for the neurotransmitter acetylcholine, it is involved in several functions including muscle and memory muscle control. It was used as a basic catalyst for aldol condensation reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Choline hydroxide is used as a precursor molecule for the neurotransmitter acetylcholine, it is involved in several functions including muscle and memory muscle control. It was used as a basic catalyst for aldol condensation reactions.

Solubility
Fully miscible in water.

Notes
Store in a cool, dry place in tightly closed container. Store away from oxidizing agents, air, acids.
TRUSTED_SUSTAINABILITY

Identifiants chimiques

CAS 123-41-1
Molecular Formula C5H15NO2
Molecular Weight (g/mol) 121.18
MDL Number MFCD00002831
InChI Key KIZQNNOULOCVDM-UHFFFAOYSA-M
Synonym choline hydroxide, gossypine, sincaline, bursine, luridine, sinkalin, sinkaline, fagine, vidine, 2-hydroxy-n,n,n-trimethylethanaminium hydroxide
PubChem CID 31255
SMILES [OH-].C[N+](C)(C)CCO

Spécification

Linear Formula HOCH2CH2N(CH3)3OH
Quantity 25 mL
UN Number UN3267
IUPAC Name (2-hydroxyethyl)trimethylazanium hydroxide
Formula Weight 121.18
Concentration or Composition (by Analyte or Components) 46% w/w aq. soln.
Chemical Name or Material Choline hydroxide
compliance-icons
Identifiant produit
  • Choline hydroxide, 46% w/w aqueous solution
Mention d'avertissement
  • Danger
Catégorie de danger
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 A
Mention de danger
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Conseils de prudence
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Informations supplémentaires
  • MIXTURE LIST-Contains : Choline hydroxide

RUO – Research Use Only

Correction du contenu d'un produit

Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.

Nom du produit

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.

Merci de nous aider à améliorer notre site web. Votre commentaire a été soumis