missing translation for 'onlineSavingsMsg'
Learn More

Choline Hydroxide in Methanol Solution (~45% w/w, stabilized with 0.5% Paraformaldehyde), TRC

Product Code. 30367548
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30367548 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30367548 Supplier Toronto Research Chemicals Supplier No. TRCC43259510G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Choline Hydroxide
CAS 123-41-1
Molecular Formula C5H15NO2
Quantity 10 g
Synonym 2-Hydroxy-N,N,N-trimethyl-ethanaminium Hydroxide (1:1), (2-Hydroxyethyl)trimethylammonium Hydroxide, (beta-Hydroxyethyl)trimethylammonium Hydroxide, Bursine, Cholinium Hydroxide, Fagine, Gossypine, Luridine, N,N,N-Trimethyl-N-(2-hydroxyethyl)ammonium Hydroxide, N,N,N-Trimethyl-N-hydroxyethylammonium Hydroxide, Sincaline, Sinkalin, Sinkaline, Trimethyl(2-hydroxyethyl)ammonium Hydroxide, Trimethyl(hydroxyethyl)ammonium Hydroxide, Vidine
InChI Formula InChI=1S/C5H14NO.H2O/c1-6(2,3)4-5-7;/h7H,4-5H2,1-3H3;1H2/q+1;/p-1
SMILES [OH-].C[N+](C)(C)CCO
IUPAC Name 2-hydroxyethyl(trimethyl)azanium;hydroxide
Molecular Weight (g/mol) 121.18
Formula Weight 121.1103
Concentration or Composition (by Analyte or Components) 45 %
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.