missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ CHPG
GREENER_CHOICE

Product Code. p-4783346
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 30001178

Brand: Tocris Bioscience™ 1049/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

mGlu5 selective agonist

A selective mGlu5 metabotropic glutamate receptor agonist, completely inactive at mGlu1a receptors expressed in CHO cells. Active in vivo.

Sodium Salt also available.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 170846-74-9
Molecular Formula C8H8ClNO3

Specifications

Quantity 10 mg
Formula Weight 201.61
Purity >99%
Molecular Weight (g/mol) 201.606
Chemical Name or Material CHPG
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.