Learn More
Macherey-Nagel™ Chromabond™ Glass Cabinet
Product Code. 10726902 Shop All Macherey-Nagel Products
Change view
Click to view available options
For Use With (Equipment):
12 Columns
16 LV or 24 Column
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
10726902 16 LV or 24 Column Each
10535561 12 Columns Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10726902 Supplier Macherey-Nagel™ Supplier No. 730174
Estimated Shipment: 27-10-2026
Add to basket
Add to basket
Glass cabinet for SPE vacuum manifolds
CHROMABOND™ vacuum manifold for simultaneous preparation of up to 12 samples. The manifold consists of a rectangular glass cabinet with a vacuum gauge and a polypropylene lid, that can hold the columns or cartridges. The replaceable valves on the lid allow individual vacuum control for each solid phase extraction column, if required. The cabinet is fitted with a variable rack with exchangeable partitions, that accept a wide variety of vessels like test tubes, measuring flasks, scintillation vials, autosampler vials, plastic vials and many more. The vacuum in the chamber can be adjusted with the control valve and read from the gauge.
Specifications
For Use With (Equipment) 16 LV or 24 Column
Type Cabinet
Material Glass
Quantity 1 Each