Learn More
Macherey-Nagel™ Chromabond™ Glass Fiber Frit
Product Code. 10347931 Shop All Macherey-Nagel Products
Change view
Click to view available options
Volume (Metric):
3 mL
6 mL
Unit Size:
Pack of 250
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Unit Size
10347931 6 mL Pack of 250
10015671 3 mL Pack of 250
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10347931 Supplier Macherey-Nagel™ Supplier No. 730192
ChemsChrom Mega Deals
Use promo code 27993
View all promotions
In Stock
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Ideal for high temperature SPE applications. Macherey-Nagel™ Chromabond™ Glass Fiber Frit are designed for individual packing of empty glass SPE columns with Chromabond™ adsorbents (glass columns, adsorbents sold separately).
Frits act as filters in solid phase extraction chromatography. They help to hold the column packing in place and facilitates sorbent packing in the column. Glass fiber columns/frits have many advantages:
  • Resistant to acids, bases and most organic solvents
  • Has low absorption for most biological macromolecules
  • Ideal for high-temperature applications
Specifications
For Use With (Application) Solid Phase Extraction
For Use With (Equipment) Glass Column
Type Frit
Volume (Metric) 6 mL
Material Glass Fiber
Quantity 250 Pack