Learn More
Macherey-Nagel™ Chromabond™ Polypropylene Tank
Product Code. 12756761 Shop All Macherey-Nagel Products
Change view
Click to view available options
For Use With (Equipment):
Vacuum Manifold 12 Columns
Unit Size:
Each
Product Code. For Use With (Equipment) Unit Size
12756761 Vacuum Manifold 12 Columns Each
1 options
Product Code. 12756761 Supplier Macherey-Nagel™ Supplier No. 730233
Estimated Shipment: 17-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
For simultaneous preparation of up to 12 samples
CHROMABOND™ vacuum manifold for simultaneous preparation of up to 12 samples. The manifold consists of a rectangular glass cabinet with a vacuum gauge and a polypropylene lid, that can hold the columns or cartridges. The replaceable valves on the lid allow individual vacuum control for each solid phase extraction column, if required. The cabinet is fitted with a variable rack with exchangeable partitions, that accept a wide variety of vessels like test tubes, measuring flasks, scintillation vials, autosampler vials, plastic vials and many more. The vacuum in the chamber can be adjusted with the control valve and read from the gauge.
Specifications
For Use With (Equipment) Vacuum Manifold 12 Columns
Type Tank
Material Polypropylene
Quantity 1 Each