missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ Chromabond™ SA Polypropylene SPE Column

Brand:  Macherey-Nagel™ 730425

Code : 50

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 12496666

  • 139.00 € / Pack of 30
Estimated Shipment: 24-06-2024
to see stock.
Add to basket

Description

Description

Specifications

Specifications

Silica
SPE
Solid Phase Extraction
Chromabond SA
Hydrophobic, p-p Interaction
500 m2/g
6 mL
500 mg
45 μm
Solid Phase Extraction Column
60 Å
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers