missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ CHROMABOND™ XTR Polypropylene SPE Columns

Product Code. 10216281 Shop All Macherey-Nagel Products
Change view
Click to view available options
Bed Weight:
250 mg
500 mg
3 g
4.5 g
8.3 g
Column Capacity (Metric):
1 mL
3 mL
15 mL
30 mL
45 mL
Unit Size:
Pack of 100
Pack of 30
Pack of 50
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Bed Weight Column Capacity (Metric) Unit Size
10216281 3 g 15 mL Pack of 30
10545371 250 mg 1 mL Pack of 100
10725952 4.5 g 30 mL Pack of 30
10690532 500 mg 3 mL Pack of 50
10286561 8.3 g 45 mL Pack of 30
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Eluents with too high alcohol contents cause an increase in volume of the aqueous phase on the CHROMABOND™ XTR. Here the column could be overloaded and the aqueous phase displaced from the column. In this case, a greater capacity column should be used.
Product Code. 10216281 Supplier Macherey-Nagel™ Supplier No. 730489
ChemsChrom Mega Deals
Use promo code 27993
View all promotions
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Eluents with too high alcohol contents cause an increase in volume of the aqueous phase on the CHROMABOND™ XTR. Here the column could be overloaded and the aqueous phase displaced from the column. In this case, a greater capacity column should be used.

Designed for consistently high quality from batch to batch. Macherey-Nagel™ CHROMABOND™ XTR Polypropylene SPE Columns include Kieselguhr phase for liquid-liquid extractions. Columns feature a large pore size and high pore volume for use with highly viscous aqueous solutions. Volumes from 1–45mL.

Polypropylene SPE columns with a base material of coarse-grained kieselguhr (also known as diatomaceous earth, hydromatrix, celite), with a large pore size and high pore volume. The pH value of aqueous solutions can be altered on the column, from 1–13, which enables elution of different compounds of a sample under optimized conditions.

Application:
For liquid-liquid extraction of highly viscous aqueous solutions without use of a separation funnel. Columns feature high water loadability without breakthrough during elution with organic solvents. They are also suited for removing small amounts of water from solvents which are not miscible with water. 

  • Physiological fluids (blood, plasma, and serum) in clinical chemistry
  • Dyes in textiles
  • Environmental and food analysis

Advantages:

  • Fast, reproducible and economical
  • Simultaneous preparation of several samples
  • No problems with phase separation—no formation of emulsions
  • High recovery rates
  • Saving of time and solvents
  • Organic solutions do not need to be dried after separation

Specifications

Column Capacity (Metric) 15 mL
Column Type Polypropylene
Bed Weight 3 g
For Use With (Application) Liquid-Liquid Extraction
Sorbent Chromabond XTR
Type Chromabond XTR Column
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.