missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ChromaCare™ LC-MS Aqueous Rinse, Probe Wash 4, Thermo Scientific™
Shop All Thermo Scientific ProductsDescription
- Suitable for cleaning sample preparation needle and drain lines on LC instrument after sampling
- Save time by using off-the-shelf blend — premade for convenience
- Helps to maintain clean sample lines and reduce sample carryover
- Packaged in 2.5L amber glass bottle
- Complementary to T001262500
Specifications
Specifications
| CAS | 75-05-8 |
| Percent Purity | 1.8 to 2.2% (Acetonitrile) |
| Concentration or Composition (by Analyte or Components) | Water with 2% Acetonitrile |
| Packaging | Amber Glass Bottle |
| Quantity | 2.5 L |
| Physical Form | Liquid |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Haben Sie Verbesserungsvorschläge?