missing translation for 'onlineSavingsMsg'
Learn More

ChromaCare™ LC-MS Biologics Flush Solution

Product Code. 15793467 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
2.5 L
Packaging:
Amber Glass Bottle
Unit Size:
2.5 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15793467 2.5 L Amber Glass Bottle 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15793467 Supplier Thermo Scientific Supplier No. MB124212
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Flush solution for cleaning blood, plasma, and urine samples from the chromatograph.

  • Suitable for flushing difficult to clean samples such as blood, plasma, and urine after vitamin D, hormone or drugs of abuse testing
  • Save time by using off-the-shelf blend — premade for convenience
  • Helps to reduce sample carryover
  • Packaged in 2.5L amber glass bottle
TRUSTED_SUSTAINABILITY

Specifications

pH 6 to 7
Boiling Point 167°F
CAS 67-64-1, 75-05-8
Synonym isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol
Percent Purity Pass Test (By GC-FID)
Grade LC-MS
Concentration or Composition (by Analyte or Components) 45% 2-Propanol, 45% Acetonitrile, 10% Acetone
Packaging Amber Glass Bottle
Quantity 2.5 L
Physical Form Liquid
compliance-icons compliance-icons
Product Identifier
  • 45% Isopropyl Alcohol + 45% Acetonitrile + 10% Acetone
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H336-May cause drowsiness or dizziness.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
Supplemental information
  • MIXTURE LIST-Contain: Acetonitrile, Isopropyl alcohol, Acetone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.