missing translation for 'onlineSavingsMsg'
Learn More

Chromacol 24mm Seals for Vial Closures

Product Code. 10726995
Change view
Click to view available options
Unit Size:
Pack of 100
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. unitSize
10726995 Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10726995 Supplier Chromacol Supplier No. 24ST3EPA

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Thermo Scientific™ Chromacol 24mm Seals are for use with EPA sampling vials.

Thermo Scientific™ Chromacol 24mm Seals are for use with EPA sampling vials.

TRUSTED_SUSTAINABILITY

Specifications

Description EPA Septa Silicone/PTFE, 24mm
Diameter (Metric) Inner 24 mm
Material PTFE, Silicone
Product Line Chromacol
Closure Size 24 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.