Learn More
Thermo Scientific™ Chromacol™ GOLD-Grade Inert Vials and Inserts
Description
Ensure sample security and integrity with inert vials that have the highest recovery rates, an ultra-low pH shift, and lowest alkali content.
Maximize your sample performance and have fewer failed assays with Thermo Scientific™ Chromacol™ GOLD-Grade Inert Glass Vials. These vials offer the most state-of-the-art technology in the industry for the analysis of critical low detection limit analytes, and when sample quality, reproducibility, security, and integrity are of upmost importance.
For easier sample handling and less risk of sample loss, the Chromacol GOLD-Grade Vials are the best-in-class choice for all chromatography users performing ultra-trace analysis.
- 8mm crimp top seal minimizes exposure between sample solvent and septum
- Neutral Borosilicate GOLD-Grade glass is a low expansion, high purity glass with an extremely low concentration of active sites to protect your analytes from unwanted interactions
- Precision-machined (glass or PTFE) sleeves allow the vials to be used in the vast majority of commercial autosamplers
Thermo Scientific™
Agilent™
Beckman™
Carlo Erba™
CTC™
Fisons™
PerkinElmer™
Shimadzu™
VWR™ (Merck)/Hitachi™
*See the Thermo Scientific Columns and Consumables Catalog for autosampler compatibility
Spécification
Spécification
| Description | 0.1mL Sci-Vi Crimp Top Vial - GOLD Grade Glass |
| Color | Clear |
| Diameter (Metric) Outer | 6 mm |
| Material | Glass |
| Volume (Metric) | 125 μL |
| Volume (Metric) Residual | <1 μL |
| Product Line | Chromacol Gold |
| Quantity | 500 |
| Closure Size | 8 mm |
| Marking Spot | No |
| Afficher plus |
En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.