Learn More
Macherey-Nagel™ Chromafix™ C18 ec Cartridge
Product Code. 10184511 Shop All Macherey-Nagel Products
Change view
Click to view available options
Bed Weight:
270 mg
950 mg
Unit Size:
Pack of 50
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Bed Weight Unit Size
10184511 950 mg Pack of 50
10348071 270 mg Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10184511 Supplier Macherey-Nagel™ Supplier No. 731806
In Stock
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Specifications
Base Material Silica
Carbon Load 14%
Bed Weight 950 mg
For Use With (Application) Non-Polar Compounds, Aflatoxins, Amphetamines, Antibiotics, Antiepileptics, Barbiturates, Caffeine, Drugs, Preservatives, Fatty Acids, Nicotine, PAHs, Pesticides, PCBs, Heavy Metals, Vitamins, Very Well Suited for Desalting of Samples
Particle Size 45 μm
Sorbent C18 ec
Type Chromafix C18 ec Cartridge
Endcapped Yes
Pore Size 60 Å
Quantity 50 Pack
Surface Area (Metric) 500 m2/g