missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ Chromafix™ C18 ec Cartridge

Product Code. 10348071 Shop All Macherey-Nagel Products
Change view
Click to view available options
Bed Weight:
270 mg
950 mg
Unit Size:
Pack of 50
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Bed Weight unitSize
10348071 270 mg Pack of 50
10184511 950 mg Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10348071 Supplier Macherey-Nagel™ Supplier No. 731804
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Specifications

Base Material Silica
Carbon Load 14%
Bed Weight 270 mg
For Use With (Application) Non-Polar Compounds, Aflatoxins, Amphetamines, Antibiotics, Antiepileptics, Barbiturates, Caffeine, Drugs, Preservatives, Fatty Acids, Nicotine, PAHs, Pesticides, PCBs, Heavy Metals, Vitamins, Very Well Suited for Desalting of Samples
Particle Size 45 μm
Sorbent C18 ec
Type Chromafix C18 ec Cartridge
Endcapped Yes
Pore Size 60 Å
Quantity 50 Pack
Surface Area (Metric) 500 m2/g
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.