Learn More
Macherey-Nagel™ Chromafix™ Dry (Na2SO4) Cartridge
Product Code. 10441282 Shop All Macherey-Nagel Products
Click to view available options
Bed Weight:
780 mg
1500 mg
2800 mg
Unit Size:
Pack of 50
Product Code. 10441282
Brand: Macherey-Nagel™ 731852
Only null left
Add to basket
Add to basket
Specifications
Base Material Dry (Na2 SO4)
Bed Weight 780 mg
For Use With (Application) Removal of Traces of Water From Organic Solutions
Type Chromafix Dry (Na2 SO4) Cartridge
Quantity 50 Pack