Learn More
Macherey-Nagel™ Chromafix™ HR-XAW Cartridge
Shop All Macherey-Nagel Products
Click to view available options
Bed Weight:
155 mg
240 mg
500 mg
Unit Size:
Pack of 50
Specifications
Specifications
| Base Material | Spherical PS/DVB Copolymer |
| Bed Weight | 155 mg |
| For Use With (Application) | Perfluorinated Surfactants, Acidic Compounds Like Sulfonates, Active Ingredients From Strongly Matrix-Contaminated Samples Like, e.g., Urine, Plasma, Serum |
| Particle Size | 45 or 85 μm |
| Sorbent | Weakly basic secondary and tertiary ammonium polymeric anion exchanger |
| Type | Chromafix HR-XAW Cartridge |
| Pore Size | 55 to 65 Å |
| Quantity | 50 Pack |
| Surface Area (Metric) | 850 m2/g |