Learn More
Macherey-Nagel™ Chromafix™ PS-Ag+ cartridges
Product Code. 10610732 Shop All Macherey-Nagel Products
Change view
Click to view available options
Quantity:
50 Pack
Unit Size:
Pack of 50
Product Code. Quantity Unit Size
10610732 50 Pack Pack of 50
1 options
Product Code. 10610732 Supplier Macherey-Nagel™ Supplier No. 731865
Estimated Shipment: 27-10-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Specifications
Base Material Strong PS/DVB Cation Exchanger, Ag+ form
For Use With (Application) Removal of Halide Ions From Water
Particle Size 100 μm
Type Chromafix PS-Ag+ Cartridge
Pore Size 100 Å
Quantity 50 Pack