Learn More
Macherey-Nagel™ Chromafix™ PS-Ba2+ cartridges
Product Code. 10402052 Shop All Macherey-Nagel Products
Change view
Click to view available options
Bed Weight:
280 mg
550 mg
Unit Size:
Pack of 50
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Bed Weight Unit Size
10402052 550 mg Pack of 50
10308601 280 mg Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10402052 Supplier Macherey-Nagel™ Supplier No. 731870
In Stock
Add to basket
Add to basket
Specifications
Base Material Strong PS/DVB Cation Exchanger, Ba2+ form
Bed Weight 550 mg
For Use With (Application) Removal of Sulfate Ions From Water
Particle Size 100 μm
Type Chromafix PS-Ba2+ Cartridge
Pore Size 100 Å
Quantity 50 Pack