missing translation for 'onlineSavingsMsg'
Learn More

Chromalite™ M Methacrylic Hydrophilic Macroporous Resins, 40 to 90 μm, Purolite™

Product Code. 17029473
Change view
Click to view available options
Chemical Name or Material:
Diethylamino Methacrylate Macroporous Weak Anion Exchanger
Epoxy Methacrylate Macroporous Affinity Resin
Iminodiacetic Acid Methacrylate Affinity Resin
Quaternary Ammonium Methacrylate Macroporous Strong Anion Exchanger
Sulfopropyl Methacrylate Macroporous Strong Cation Exchanger
Physical Form:
20% EtOH slurry
Spherical Beads
Wet
Quantity:
25 mL
100 mL
500 mL
Unit Size:
100mL
25mL
500mL
15 product options available for selection
Product selection table with 15 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Chemical Name or Material Physical Form Quantity unitSize
17029473 Diethylamino Methacrylate Macroporous Weak Anion Exchanger 20% EtOH slurry 100 mL 100mL
17049473 Diethylamino Methacrylate Macroporous Weak Anion Exchanger 20% EtOH slurry 25 mL 25mL
17039473 Diethylamino Methacrylate Macroporous Weak Anion Exchanger 20% EtOH slurry 500 mL 500mL
17099443 Epoxy Methacrylate Macroporous Affinity Resin Wet 100 mL 100mL
17089443 Epoxy Methacrylate Macroporous Affinity Resin Wet 25 mL 25mL
17009453 Epoxy Methacrylate Macroporous Affinity Resin Wet 500 mL 500mL
17079453 Iminodiacetic Acid Methacrylate Affinity Resin Spherical Beads 100 mL 100mL
17099453 Iminodiacetic Acid Methacrylate Affinity Resin Spherical Beads 25 mL 25mL
17089453 Iminodiacetic Acid Methacrylate Affinity Resin Spherical Beads 500 mL 500mL
17069463 Quaternary Ammonium Methacrylate Macroporous Strong Anion Exchanger Spherical Beads 100 mL 100mL
17089463 Quaternary Ammonium Methacrylate Macroporous Strong Anion Exchanger Spherical Beads 25 mL 25mL
17079463 Quaternary Ammonium Methacrylate Macroporous Strong Anion Exchanger Spherical Beads 500 mL 500mL
17019483 Sulfopropyl Methacrylate Macroporous Strong Cation Exchanger Spherical Beads 100 mL 100mL
17039483 Sulfopropyl Methacrylate Macroporous Strong Cation Exchanger Spherical Beads 25 mL 25mL
17029483 Sulfopropyl Methacrylate Macroporous Strong Cation Exchanger Spherical Beads 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 15 options available.
15 options
This item is not returnable. View return policy
Product Code. 17029473 Supplier Chromalite Supplier No. LS04311164

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chromalite™ M resins are hydrophilic, highly porous with high mechanical stability, based on a rigid methacrylic backbone. Optimal for protein purification and separation.

Hydrophilic, macroporous resins based on a methacrylic backbone.

  • Ion exchange chromatography
  • Typical porosity of 1000Å suitable for protein purification (up to 250 kDa)
  • Suitable for large-scale applications
  • Excellent pressure-flow properties,designed for high performance, stability and reliability
  • Purification of peptides obtained by fermentation
  • High mechanical stability
  • Uniform particle size
  • Stable to CIP (up to 1M NaOH)
  • Negligible swelling
  • Compliant with ResAP(2004) 3
  • Halal, Kosher and TSE/BSE/GMO free

Specifications

Chemical Name or Material Diethylamino Methacrylate Macroporous Weak Anion Exchanger
Physical Form 20% EtOH slurry
Particle Size 40 to 90 μm
Pore Size 1000 Å
Quantity 100 mL
Product Line Chromalite™ MDEA/F
Functionality/Interaction Type Diethylamino Methacrylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.