missing translation for 'onlineSavingsMsg'
Learn More

Chroman-6-carbaldehyde, 95%, Thermo Scientific™

Product Code. 10316764
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10316764 1 g Amber Glass Bottle 1g
10113514 250 mg Amber Glass Bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10316764 Supplier Thermo Scientific Maybridge Supplier No. CC32804DA
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 55745-97-6
Molecular Formula C10H10O2
Molecular Weight (g/mol) 162.188
MDL Number MFCD08146589
InChI Key YIHDTNNFJDJYRG-UHFFFAOYSA-N
Synonym chroman-6-carbaldehyde, chromane-6-carbaldehyde, 3,4-dihydro-2h-1-benzopyran-6-carbaldehyde, chroman-6-carboxaldehyde, 3,4-dihydro-2h-chromene-6-carboxaldehyde, 2h-1-benzopyran-6-carboxaldehyde, 3,4-dihydro
PubChem CID 6504215
IUPAC Name 3,4-dihydro-2H-chromene-6-carbaldehyde
SMILES C1CC2=C(C=CC(=C2)C=O)OC1

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 162.19
Percent Purity 95%
Chemical Name or Material Chroman-6-carbaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.