missing translation for 'onlineSavingsMsg'
Learn More

Chroman-6-carbonyl chloride, 95%, Thermo Scientific™

Product Code. 10173074
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10173074 1 g Amber Glass Bottle 1g
10113034 250 mg Amber Glass Bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10173074 Supplier Thermo Scientific Maybridge Supplier No. CC32802DA
 Restricted Item
Estimated Shipment: 05-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 55746-03-7
Molecular Formula C10H9ClO2
Molecular Weight (g/mol) 196.63
MDL Number MFCD12198108
InChI Key DKSNWGZAMLPCLZ-UHFFFAOYSA-N
Synonym chroman-6-carbonyl chloride, 3,4-dihydro-2h-1-benzopyran-6-carbonyl chloride, chromane-6-carbonyl chloride, 2h-1-benzopyran-6-carbonyl chloride,3,4-dihydro
PubChem CID 10442680
IUPAC Name 3,4-dihydro-2H-chromene-6-carbonyl chloride
SMILES C1CC2=C(C=CC(=C2)C(=O)Cl)OC1

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 196.63
Percent Purity 95%
Chemical Name or Material Chroman-6-carbonyl chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.