missing translation for 'onlineSavingsMsg'
Learn More

Chroman-6-carboxylic acid, 97%, Thermo Scientific™

Product Code. 10708255
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10708255 250 mg Amber Glass Bottle 250mg
10285264 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10708255 Supplier Thermo Scientific Maybridge Supplier No. CC32801CB
 Restricted Item
Estimated Shipment: 28-10-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 103203-84-5
Molecular Formula C10H10O3
Molecular Weight (g/mol) 178.19
MDL Number MFCD08146590
InChI Key IFKANGOXGBPILW-UHFFFAOYSA-N
Synonym chroman-6-carboxylic acid, 3,4-dihydro-2h-1-benzopyran-6-carboxylic acid, chromane-6-carboxylic acid, chroman-6-carboxylicacid, 2h-1-benzopyran-6-carboxylicacid, 3,4-dihydro, 6-chromanecarboxylic acid, acmc-1c8ke, 3,4-dihydro-2h-benzopyran-6-carboxylic acid
PubChem CID 6504216
IUPAC Name 3,4-dihydro-2H-chromene-6-carboxylic acid
SMILES OC(=O)C1=CC2=C(OCCC2)C=C1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 178.19
Percent Purity 97%
Chemical Name or Material Chroman-6-carboxylic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.