missing translation for 'onlineSavingsMsg'
Learn More

Chroman-8-carboxylic acid, 97%, Thermo Scientific™

Product Code. 10505194
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10505194 1 g Amber Glass Bottle 1g
10266004 250 mg Amber Glass Bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10505194 Supplier Thermo Scientific Maybridge Supplier No. CC76101DA
 Restricted Item
Estimated Shipment: 11-02-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 31457-16-6
Molecular Formula C10H10O3
Molecular Weight (g/mol) 178.187
MDL Number MFCD12198122
InChI Key LOFOWPRKKPHPDW-UHFFFAOYSA-N
Synonym chroman-8-carboxylic acid, 3,4-dihydro-2h-1-benzopyran-8-carboxylic acid, chromane-8-carboxylic acid
PubChem CID 12701138
IUPAC Name 3,4-dihydro-2H-chromene-8-carboxylic acid
SMILES C1CC2=CC=CC(=C2OC1)C(=O)O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 178.19
Percent Purity 97%
Chemical Name or Material Chroman-8-carboxylic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.