missing translation for 'onlineSavingsMsg'
Learn More

Chromate, Quant™ Test Strips

Product Code. 15431127
Change view
Click to view available options
Quantity:
1 Kit
Unit Size:
Each
Product Code. Quantity unitSize
15431127 1 Kit Each
1 options
This item is not returnable. View return policy

Product Code. 15431127

Brand: Thermo Scientific Alfa Aesar 023274.QA

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

 

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Quantity 1 Kit
Range Measures 3 to 1000ppm CrO4-2 in aqueous solution
Type Chromate test strip
What comes with the Chromate, Quant Test Strips?

When you purchase the Chromate, Quant Test Strips, you will receive the following:

  • 1 Tube containing 100 test strips
  • 1 bottle of reagent CrO4-1
  • 1 test vessel with 5 mL mark

What are the graduations of the Chromate, Quant Test Strips?

The graduations are as follows: 3 – 10 – 30 – 100 mg/L CrO42-

What instructions should I use for the Chromate, Quant Test Strips?

We recommend the following instructions for the Chromate, Quant Test Strips:
1. Rinse the test vessel several times with the sample.
2. Fill the test vessel to the 5 mL mark with sample.
3. Add Reagent CrO4-1 dropwise until the pH of the solution is below 1. This can be checked with pH test strips (not included).
4. Immerse the reaction zone of the test strip in the sample for 1 second.
5. Allow excess liquid to run off via the long edge of the strip onto an absorbent paper towel.
6. After 15 seconds, compare the color of the reaction zone to the color field on the label.
7. Read off the corresponding result.

compliance-icons
Product Identifier
  • Chromate, Quant Test Strips
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H290-May be corrosive to metals.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : Sulphuric acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.