missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ChromaTide™ Alexa Fluor™ 594-5-dUTP

Product Code. 10143622
Change view
Click to view available options
Quantity:
25 μL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10143622 25 μL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10143622 Supplier Invitrogen™ Supplier No. C11400
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

ChromaTide™ Alexa Fluor™ 594-5-dUTP

Molecular Probes™ ChromaTide™ dye labeled dUTP, OBEA-dCTP, and UTP nucleotides can be used to synthesize labeled DNA probes without the need for hazardous and expensive radioisotope-labeled nucleotides. These nucleotides can be incorporated using standard molecular biology techniques; labeled probes can then be used in in situ hybridization, microarray, or blotting protocols. ChromaTide™ dye labeled nucleotides are available in different fluorescent colors to facilitate multicolor analysis.

ChromaTide™ Labeled Nucleotides Specifications:
  • Ex/Em of dye: Alexa Fluor™ 594-5-dUTP (590/615 nm)
  • Length of alkynylamino linker: 5 atoms

    Methods for Incorporating ChromaTide™ Nucleotides Into Probes
  • Nick translation
  • Random primer labeling
  • End-labeling with terminal deoxynucleotidyl transferase
  • Reverse transcription
  • PCR amplification

    See Methods for Enzymatic Incorporation of ChromaTide™ dUTPs for specific guidelines for each of these methods.

    Alexa Fluor™ and BODIPY™ Fluorescent Dyes Make Excellent Probes
    Probes made with labeled nucleotides can be used for multicolor techniques such as in situ hybridization and hybridization to arrays. Our proprietary BODIPY™ and Alexa Fluor™ dye conjugates are exceptionally bright, photostable, and essentially pH insensitive. The narrow emission profile of the BODIPY™ dyes helps ensure minimal spectral overlap. The Alexa Fluor™ dyes are highly water soluble, as are DNA probes made from them, making them the labels of choice for fluorescence in situ hybridization.

    Long Linkers Improve Performance
    The ChromaTide™ dUTP and UTP nucleotides are modified at the C-5 position of uridine via a unique alkynylamino linker, which provides a spacer between the nucleotide and the dye to reduce interactions between them. The number in the product name, e.g., the “12” in fluorescein-12-dUTP, indicates the net length of the spacer, in atoms. These spacers result in brighter conjugates and increased hapten accessibility for secondary detection reagents.

    For complete listing of our ChromaTide™ Reagents: Molecular Probes ChromaTide™ and aha labeled nucleotides—Table 8.5.
    For additional information on these labeling reagents, read Labeling Oligonucleotides and Nucleic Acids—Section 8.2 in the Molecular Probes™ Handbook.

    For Research Use Only. Not intended for human or animal therapeutic or diagnostic use.
  • TRUSTED_SUSTAINABILITY

    Spezifikation

    Content And Storage Store in freezer (-5 to -30°C) and protect from light.
    Labeling Method Direct Labeling
    Shipping Condition Dry Ice
    Concentration 1 mM
    Label or Dye Alexa Fluor™ 594
    Quantity 25 μL
    Product Line Alexa Fluor, ChromaTide

    For Research Use Only. Not for use in diagnostic procedures.

    Name des Produkts
    Wählen Sie ein Thema

    Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.