Learn More
Fisherbrand™ Chromatography Columns
Product Code. 15379064 Shop All Fisherbrand Products
Change view
Click to view available options
Pore Size:
3
4
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Pore Size unitSize
15379064 3 Each
15399074 4 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15379064
Estimated Shipment: 24-08-2026
Add to basket
Add to basket
Increase sample throughput and laboratory productivity with Fisherbrand™ Chromatography Columns for all compound separation needs.
  • For separation of compounds
  • With sintered glass fritted disk
  • With PTFE keys
Specifications
Quantity 1
Type Chromatography Column
Pore Size 3
Length (Metric) 100 mm