Learn More
Macherey-Nagel™ Chrombond™ RS 120 NH2 Polypropylene Flash Column
Product Code. 12717760 Shop All Macherey-Nagel Products
Change view
Click to view available options
Length (Metric):
255 mm
Unit Size:
Each
Product Code. Length (Metric) unitSize
12717760 255 mm Each
1 options
Product Code. 12717760 Supplier Macherey-Nagel™ Supplier No. 732825
Estimated Shipment: 18-08-2026
Add to basket
Add to basket
Specifications
Length (Metric) 255 mm
Material Polypropylene
Type Flash Column
Column Type Flash Column
For Use With (Equipment) Directly Used in The Teledyne Isco Companion, Rf