Learn More
Macherey-Nagel™ Chrombond™ RS 80 C18 ec Polypropylene Flash Column
Product Code. 12738332 Shop All Macherey-Nagel Products
Change view
Click to view available options
Length (Metric):
240 mm
Unit Size:
Each
Product Code. Length (Metric) Unit Size
12738332 240 mm Each
1 options
Product Code. 12738332 Supplier Macherey-Nagel™ Supplier No. 732814
Big Savings for Better Separation
Use promo code 27992
View all promotions
In Stock
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Specifications
Length (Metric) 240 mm
Material Polypropylene
Type Flash Column
Column Type Flash Column
For Use With (Equipment) Directly Used in The Teledyne Isco Companion, Rf