Learn More
Thermo Scientific™ Chromeleon™ CDS Single Edition (SE) License
Product Code. 31903479 Shop All Thermo Scientific Products
Change view
Click to view available options
For Use With (Equipment):
Chromeleon CDS
Unit Size:
Each
Product Code. For Use With (Equipment) Unit Size
31903479 Chromeleon CDS Each
1 options
Product Code. 31903479 Supplier Thermo Scientific™ Supplier No. 7100.0108
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
missing translation for 'validMainframeMsg'
Single Edition (SE) Chromeleon™ CDS license enabling control of one Thermo Scientific instrument with data acquisition, processing, and reporting capabilities.
  • Chromeleon™ CDS Single Edition (SE) license for control of one Thermo Scientific instrument
  • Includes one client enabling local or remote instrument control
  • Supports instrument control, data acquisition, processing, and reporting
  • Includes Instrument Controller and Class 1 instrument license
  • Includes Data Client, Instrument Operation, and Report Designer Pro
  • Includes Fraction Collection functionality
  • SE license is a stand-alone license and cannot be added to an existing Chromeleon installation
  • Non-Thermo Scientific instruments cannot be added to this configuration
  • License must be ordered with the appropriate license key or license code depending on Chromeleon version
TRUSTED_SUSTAINABILITY
Specifications
Type Data System
For Use With (Equipment) Chromeleon CDS