Learn More
Thermo Scientific Chromeleon™ Enterprise Client
Product Code. 31906283 Shop All Thermo Scientific Products
Change view
Click to view available options
Operating System:
Windows 11
Unit Size:
Each
Product Code. Operating System Unit Size
31906283 Windows 11 Each
1 options
Product Code. 31906283 Supplier Thermo Scientific Supplier No. 7200.0300
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
missing translation for 'validMainframeMsg'
The Enterprise Client license provides tools for post-acquisition data processing, compliance, and reporting.
Includes:
  • Data Client for data processing
  • Compliance tools, enabling user management, object audit trails, versioning and electronic signatures
  • Instrument Operation (both locally and remotely)
  • Virtual Column Complete, enabling modeling of both isocratic and linear gradient separations
  • Report Designer Pro for creation and/or charting of custom Excel-style formulas.
TRUSTED_SUSTAINABILITY
Specifications
Type Chromatography Software
For Use With (Equipment) Chromeleon CDS
Certifications/Compliance 21 CFR
Product Line Chromeleon 7
Operating System Windows 11
Quantity 1 Ea.