missing translation for 'onlineSavingsMsg'
Learn More

Chromium dioxide, TRC

Product Code. 30354815
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30354815 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30354815 Supplier Toronto Research Chemicals Supplier No. TRCC27411025G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Chromium dioxide
CAS 12018-01-8
Molecular Formula CrO2
Quantity 25 g
Synonym Chromium oxide (CrO2), Amdry 6415, Chromium dioxide, Chromium dioxide (CrO2), Chromium oxide, Chromium(IV) oxide, Magtrieve
InChI Formula InChI=1S/Cr.2O
SMILES O=[Cr]=O
IUPAC Name dioxochromium
Molecular Weight (g/mol) 83.99
Formula Weight 83.9303
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.