missing translation for 'onlineSavingsMsg'
Learn More

Chromium(II) chloride, 99.9% (metals basis)

Product Code. 15462887
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15462887 1 g 1g
15472887 5 g 5g
15482887 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15462887 Supplier Thermo Scientific Alfa Aesar Supplier No. 047176.03
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 10049-05-5

Chromium(II) chloride is used in the condensation of aldehydes with trisubstituted chloroalkenes as well as in the synthesis of a highly active precatalyst for ethylene oligimerization. Further uses of chromium(II) chloride include the conversion of terminal olefins into trans-iodocyclopropanes (in conjunction with iodoform and tetraethyl ethylenediamine) with excellent results; generating bis(indenyl)chromium dimers and other various related products.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10049-05-5
MDL Number MFCD00010947
Synonym Chromous chloride

Specifications

Melting Point 824°C
Color Gray
Physical Form Powder
Quantity 1 g
Linear Formula CrCl2
UN Number UN3260
Merck Index 14,2244
Solubility Information Soluble in water. Insoluble in alcohol and ether.
Formula Weight 122.9
Percent Purity 99.9%
Odor Odorless
Sensitivity Air and moisture sensitive
Density 2.878 g/mL
Assay (metals basis)
Chemical Name or Material Chromium(II) chloride
Show More Show Less
compliance-icons
Product Identifier
  • Chromium(II) chloride, anhydrous
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Chromic(II) chloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.