missing translation for 'onlineSavingsMsg'
Learn More

Chromium(II) chloride, anhydrous, 97%

Product Code. 11353486
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11353486 100 g 100g
11333486 5 g 5g
11343486 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11353486 Supplier Thermo Scientific Alfa Aesar Supplier No. L14342.22
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 10049-05-5 | CrCl2

Chromium(II) chloride is used as a precursor to other inorganic and organometallic chromium complexes. It is a reagent in the Nozaki-Hiyama-Kishi reaction. It is also used in the Takai olefination to prepare vinyl iodides from aldehydes in the presence of iodoform.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Chromium(II) chloride is used as a precursor to other inorganic and organometallic chromium complexes. It is a reagent in the Nozaki-Hiyama-Kishi reaction. It is also used in the Takai olefination to prepare vinyl iodides from aldehydes in the presence of iodoform.

Solubility
Soluble in water. Insoluble in alcohol and ether.

Notes
Hygroscopic. Incompatible with strong oxidizing agents, water and active metals.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10049-05-5
Molecular Formula CrCl2
MDL Number MFCD00010947
Synonym Chromous chloride

Specifications

Melting Point 824°C
Quantity 100 g
Odor Odorless
Sensitivity Air and moisture sensitive
Density 2.878
UN Number UN3260
Merck Index 14,2244
Solubility Information Soluble in water. Insoluble in alcohol and ether.
Formula Weight 122.9
Percent Purity 97%
Chemical Name or Material Chromium(II) chloride, anhydrous
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.