missing translation for 'onlineSavingsMsg'
Learn More

Chromium(III) 2,4-pentanedionate, 97%

Product Code. 11397377
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11397377 500 g 500g
11387377 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11397377 Supplier Thermo Scientific Alfa Aesar Supplier No. 012538.36
 Restricted Item
Estimated Shipment: 14-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Catalyst in the oxidation of methacrylic esters by hydrogen peroxide, MOCVD | CAS: 21679-31-2 | C15H21CrO6 | 349.32 g/mol

Catalyst in the oxidation of methacrylic esters by hydrogen peroxide, MOCVDChromium(III) 2,4-pentanedionate is used in the reduction of detonation of nitromethane. It is also used in NMR spectroscopy as a relaxation agent due to its solubility in nonpolar organic solvents and its paramagnetism. Further, it is involved in the modification of the surface properties of solid polyurethanes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 21679-31-2
Molecular Formula C15H21CrO6
Molecular Weight (g/mol) 349.32
MDL Number MFCD00000015 MFCD00000015
InChI Key JWORPXLMBPOPPU-LNTINUHCSA-K
Synonym chromium iii acetylacetonate, dsstox_cid_30421, dsstox_gsid_51863
PubChem CID 91759531
IUPAC Name chromium;(Z)-4-oxoniumylidenepent-2-en-2-olate
SMILES [Cr+3].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O

Specifications

Melting Point 216°C
Density 1.35 g/mL
Boiling Point 340°C
Quantity 500 g
Solubility Information Soluble in toluene,acetic acid and non-polar organic solvents; Insoluble in water.
Formula Weight 349.33
Percent Purity 97%
Physical Form Crystalline
Chemical Name or Material Chromium(III) 2,4-pentanedionate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.