missing translation for 'onlineSavingsMsg'
Learn More

Chromium(III) acetylacetonate, 97%

Product Code. 10521641 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass Bottle
Plastic bottle
Unit Size:
100g
500g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10521641 5 g Glass Bottle 5g
10750701 100 g Plastic bottle 100g
10279873 500 g Plastic bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10521641 Supplier Thermo Scientific Acros Supplier No. 192130050
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 21679-31-2 | C15H21CrO6 | 349.32 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 21679-31-2
Molecular Formula C15H21CrO6
Molecular Weight (g/mol) 349.32
MDL Number MFCD00000015 MFCD00000015
InChI Key JWORPXLMBPOPPU-LNTINUHCSA-K
Synonym chromium iii acetylacetonate, dsstox_cid_30421, dsstox_gsid_51863
PubChem CID 91759531
IUPAC Name chromium;(Z)-4-oxoniumylidenepent-2-en-2-olate
SMILES [Cr+3].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O

Specifications

Melting Point 212°C
Color Maroon to Purple
Boiling Point 340°C
Infrared Spectrum Authentic
Assay Percent Range 14.4 to 15.3% (Cr)
Packaging Glass Bottle
Linear Formula (CH3COCH=C(CH3)O)3Cr
Quantity 5 g
Solubility Information Solubility in water: 11g/L (20°C). Other solubilities: 2.20g/100g alcohol (20°C),30.85g/100g benzene (20°C),insoluble in petroleum ether
Formula Weight 349.32
Percent Purity 97%
Physical Form Crystalline Powder, Crystals or Chunks
Chemical Name or Material Chromium(III) acetylacetonate
Show More Show Less
compliance-icons
Nombre del producto
  • Chromium(III) acetylacetonate
Palabra de advertencia
  • Warning
Clasificación
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Indicaciones de peligro
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Consejos de prudencia
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.