missing translation for 'onlineSavingsMsg'
Learn More

Chromium(III) Chloride Hexahydrate, MP Biomedicals

Product Code. 11470582 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
Product Code. Quantity unitSize
11470582 100 g 100g
11480582 500 g 500g
2 options
This item is not returnable. View return policy

Product Code. 11470582

Brand: MP Biomedicals 0219403380

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 10060-12-5
Molecular Formula Cl3CrH12O6
Molecular Weight (g/mol) 266.44
MDL Number MFCD00149660
InChI Key LJAOOBNHPFKCDR-UHFFFAOYSA-K
Synonym chromium iii chloride hexahydrate, chromic chloride hexahydrate, hexaaquachromium chloride, chromium trichloride hexahydrate, trichlorochromium hexahydrate, chromium chloride hexahydrate, hexaaquachromium iii chloride, chrom iii-chlorid, chromium 3+ chloride hexahydrate, chlorid chromity hexahydrat czech
PubChem CID 104957
ChEBI CHEBI:53442
IUPAC Name chromium(3+) hexahydrate trichloride
SMILES O.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Cr+3]

Specifications

Quantity 100 g
Formula Weight 266.4
Chemical Name or Material Chromium(III) Chloride Hexahydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.