missing translation for 'onlineSavingsMsg'
Learn More

Chromium(III) Chloride Hexahydrate, TRC

Product Code. 30362090
Change view
Click to view available options
Quantity:
50 g
100 g
Unit Size:
100g
50g
Product Code. Quantity unitSize
30362090 50 g 50g
30359751 100 g 100g
2 options
This item is not returnable. View return policy

Product Code. 30362090

Brand: Toronto Research Chemicals TRCC43262050G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Chromium(III) Chloride Hexahydrate
CAS 10060-12-5
Molecular Formula Cl3CrH12O6
Quantity 50 g
Synonym Chromium chloride hexahydrate, Chromic chloride hexahydrate, Chromium chloride hexahydrate, Chromium trichloride hexahydrate, Chromium(III) chloride hexahydrate, Hexaaquachromium chloride, Hexaaquachromium(III) chloride
InChI Formula InChI=1S/3ClH.Cr.6H2O/h3*1H;;6*1H2/q;;;+3;;;;;;/p-3
SMILES O.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Cr+3]
IUPAC Name chromium(3+);trichloride;hexahydrate
Molecular Weight (g/mol) 266.45
Formula Weight 264.9105
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.