missing translation for 'onlineSavingsMsg'
Learn More

Chromium(III) oxide, 99%, amorphous powder

Product Code. 10591251 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Glass bottle
Plastic Bottle
Unit Size:
2.5kg
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10591251 500 g Glass bottle 500g
11954141 2.5 kg Plastic Bottle 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10591251 Supplier Thermo Scientific Acros Supplier No. 192085000
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1308-38-9 | Cr2O3 | 151.99 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1308-38-9
Molecular Formula Cr2O3
Molecular Weight (g/mol) 151.99
MDL Number MFCD00010949
InChI Key UOUJSJZBMCDAEU-UHFFFAOYSA-N
Synonym chromium iii oxide, chromia, dichromium trioxide, chromium sesquioxide, chrome green, green chromium oxide, chromium 3+ oxide, green cinnabar, chrome ochre, chrome oxide
PubChem CID 517277
ChEBI CHEBI:48242
IUPAC Name dichromium(3+) trioxidandiide
SMILES [O--].[O--].[O--].[Cr+3].[Cr+3]

Specifications

Boiling Point 4000°C
Melting Point 2435°C
Color Green
pH 5 to 7
Physical Form Amorphous Powder
Quantity 500 g
Assay Percent Range 98.5% min
Linear Formula Cr2O3
Fieser 03,38; 13,338
Merck Index 15,2238
Solubility Information Solubility in water: insoluble in water. Other solubilities: slightly soluble in acids and alkalies,practically insoluble in alcohol and acetone
Formula Weight 151.99
Percent Purity 99%
Packaging Glass bottle
Specific Gravity 5.21
Density 5.2100g/mL
Chemical Name or Material Chromium(III) oxide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.