missing translation for 'onlineSavingsMsg'
Learn More

Chromium(III) Potassium Sulfate, 98+%, Extra Pure, SLR, Fisher Chemical

Product Code. 12307143 Shop All Fisher Chemical Products
Change view
Click to view available options
500g, Plastic powder jar
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
12307143 500g, Plastic powder jar
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12307143 Supplier Fisher Chemical Supplier No. C/5720/53
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

Melting Point 89°C
CAS 7788-99-0
Color Purple
pH 3
Physical Form Solid
Quantity 500 g
Molecular Formula CrKO8S2·12H2O
MDL Number 149917
Formula Weight 499.39g/mol
Packaging Plastic powder jar
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.